Assay Details
| Gene Symbol: PNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079490.1 | NP_001072958.1 | 2 - 3 | 412 | 85 | |
| GenBank mRNA | AJ851486.1 | - | 2 - 3 | 412 | 85 | |
More Information
Set Membership: |
|

PNO1
Genomic Map