Assay Details
| Gene Symbol: FAM26E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079492.1 | NP_001072960.1 | 1 - 2 | 697 | 93 | |
| GenBank mRNA | AJ851418.1 | - | 1 - 2 | 697 | 93 | |
| CR407182.1 | - | 2 - 3 | 466 | 93 | ||
More Information
Set Membership: |
|

FAM26E
Genomic Map