Assay Details
Gene Symbol: RPL37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079493.2 | NP_001072961.1 | 3 - 4 | 228 | 100 | |
GenBank mRNA | CD764696.1 | - | 3 - 4 | 228 | 100 | |
CO504740.1 | - | 2 - 3 | 270 | 100 | ||
D10731.1 | - | 2 - 3 | 149 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
