Assay Details
| Gene Symbol: SUMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079498.1 | NP_001072966.1 | 2 - 3 | 234 | 108 | |
| GenBank mRNA | AJ721083.1 | - | 2 - 3 | 234 | 108 | |
More Information
Set Membership: |
|

SUMO3
Genomic Map