Assay Details
| Gene Symbol: MINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079723.1 | NP_001073191.1 | 5 - 6 | 824 | 71 | |
| GenBank mRNA | AJ720509.1 | - | 5 - 6 | 824 | 71 | |
More Information
Set Membership: |
|

MINA
Genomic Map