Assay Details
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079728.1 | NP_001073196.1 | 2 - 3 | 160 | 79 | |
XM_025149562.1 | XP_025005330.1 | 2 - 3 | 172 | 79 | ||
GenBank mRNA | AJ720025.1 | - | 2 - 3 | 160 | 79 | |
EU477531.1 | - | 1 - 2 | 107 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
