Assay Details
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079728.1 | NP_001073196.1 | 4 - 5 | 436 | 117 | |
XM_025149562.1 | XP_025005330.1 | 4 - 5 | 448 | 117 | ||
GenBank mRNA | AJ720025.1 | - | 4 - 5 | 436 | 117 | |
EU477531.1 | - | 3 - 4 | 383 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
