Assay Details
| Gene Symbol: RBMXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079728.1 | NP_001073196.1 | 5 - 6 | 562 | 71 | |
| XM_025149562.1 | XP_025005330.1 | 5 - 6 | 574 | 71 | ||
| GenBank mRNA | AJ720025.1 | - | 5 - 6 | 562 | 71 | |
| EU477531.1 | - | 4 - 5 | 509 | 71 | ||
More Information
Set Membership: |
|

RBMXL1
Genomic Map