Assay Details
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079728.1 | NP_001073196.1 | 7 - 8 | 795 | 87 | |
GenBank mRNA | AJ720025.1 | - | 7 - 8 | 795 | 87 | |
EU477531.1 | - | 6 - 7 | 742 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
