Assay Details
Gene Symbol: UCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080212.1 | NP_001073681.1 | 7 - 8 | 528 | 85 | |
XM_025149756.1 | XP_025005524.1 | 6 - 7 | 743 | 85 | ||
GenBank mRNA | AJ621938.1 | - | 7 - 8 | 528 | 85 | |
BX950489.2 | - | 7 - 8 | 606 | 85 | ||
CR387474.1 | - | 5 - 6 | 411 | 85 | ||
CR407084.1 | - | 5 - 6 | 407 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
