Assay Details
Gene Symbol: VAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080869.1 | NP_001074338.1 | 3 - 4 | 392 | 98 | |
XM_015282511.2 | XP_015137997.1 | 3 - 4 | 380 | 98 | ||
XM_025147349.1 | XP_025003117.1 | 3 - 4 | 644 | 98 | ||
GenBank mRNA | AJ851481.1 | - | 3 - 4 | 392 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
