Assay Details
| Gene Symbol: ARHGDIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080871.1 | NP_001074340.1 | 2 - 3 | 294 | 59 | |
| XM_015295424.2 | XP_015150910.1 | 2 - 3 | 288 | 59 | ||
| GenBank mRNA | AJ719303.1 | - | 2 - 3 | 294 | 59 | |
More Information
Set Membership: |
|

ARHGDIA
Genomic Map