Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080880.1 | NP_001074349.1 | 2 - 3 | 306 | 73 | |
| XM_015279150.2 | XP_015134636.1 | 2 - 3 | 304 | 73 | ||
| GenBank mRNA | AJ719553.1 | - | 2 - 3 | 306 | 73 | |
| AJ719583.1 | - | 1 - 2 | 131 | 73 | ||
More Information
Set Membership: |
|

TMEM170A
Genomic Map