Assay Details
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080892.1 | NP_001074361.1 | 4 - 5 | 506 | 97 | |
XM_015273064.2 | XP_015128550.1 | 4 - 5 | 644 | 97 | ||
XM_015273066.2 | XP_015128552.1 | 4 - 5 | 694 | 97 | ||
GenBank mRNA | AJ721103.1 | - | 4 - 5 | 506 | 97 | |
CR389489.1 | - | 1 - 2 | 132 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
