Assay Details
| Gene Symbol: RPS3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082417.2 | NP_001075886.1 | 3 - 4 | 392 | 61 | |
| GenBank mRNA | AJ737079.1 | - | 3 - 4 | 408 | 61 | |
| BU410973.1 | - | 1 - 2 | 166 | 61 | ||
| CF250059.1 | - | 3 - 4 | 394 | 61 | ||
More Information
Set Membership: |
|

RPS3A
Genomic Map