Assay Details
| Gene Symbol: RPS3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082417.2 | NP_001075886.1 | 4 - 5 | 602 | 79 | |
| GenBank mRNA | AJ737079.1 | - | 2 - 3 | 198 | 79 | |
| BU410973.1 | - | 2 - 3 | 376 | 79 | ||
| CF250059.1 | - | 4 - 5 | 604 | 79 | ||
More Information
Set Membership: |
|

RPS3A
Genomic Map