Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097532.3 | NP_001091001.1 | 2 - 3 | 248 | 80 | |
| GenBank mRNA | AJ720510.1 | - | 2 - 3 | 204 | 80 | |
| BX930878.2 | - | 2 - 3 | 150 | 80 | ||
| BX932133.2 | - | 2 - 3 | 196 | 80 | ||
More Information
Set Membership: |
|

ATP6V0E1
Genomic Map