Assay Details
| Gene Symbol: DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099353.2 | NP_001092823.1 | 3 - 4 | 679 | 78 | |
| GenBank mRNA | BX931224.2 | - | 3 - 4 | 618 | 78 | |
| HM545127.1 | - | 3 - 4 | 601 | 78 | ||
More Information
Set Membership: |
|

DMA
Genomic Map