Assay Details
| Gene Symbol: NSMCE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105064.1 | NP_001098534.1 | 522 | 98 | ||
| GenBank mRNA | BX930943.2 | - | 386 | 98 | ||
| BX932334.2 | - | 522 | 98 | |||
| BX934453.2 | - | 98 | ||||
| CR354280.1 | - | 427 | 98 | |||
More Information
Set Membership: |
|

NSMCE3
Genomic Map