Assay Details
| Gene Symbol: MSANTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109786.1 | NP_001103256.1 | 3 - 4 | 837 | 120 | |
| XM_015298049.2 | XP_015153535.1 | 4 - 5 | 908 | 120 | ||
| XM_015298050.2 | XP_015153536.1 | 4 - 5 | 905 | 120 | ||
| XM_015298051.1 | XP_015153537.1 | 3 - 4 | 5890 | 120 | ||
| XM_015298053.1 | XP_015153539.1 | 4 - 5 | 5935 | 120 | ||
| XM_015298054.1 | XP_015153540.1 | 4 - 5 | 5932 | 120 | ||
| XM_025143212.1 | XP_024998980.1 | 4 - 5 | 5815 | 120 | ||
| GenBank mRNA | AJ721009.1 | - | 3 - 4 | 837 | 120 | |
More Information
Set Membership: |
|

MSANTD2
Genomic Map