Assay Details
Gene Symbol: FGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109787.1 | NP_001103257.1 | 11 - 12 | 1475 | 101 | |
GenBank mRNA | X67786.1 | - | 1475 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
