Assay Details
| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204143.3 | NP_989474.1 | 2 - 3 | 529 | 65 | |
| XM_025142476.1 | XP_024998244.1 | 3 - 4 | 2126 | 65 | ||
| GenBank mRNA | AF401291.2 | - | 2 - 3 | 529 | 65 | |
More Information
Set Membership: |
|

AGMAT
Genomic Map