Assay Details
| Gene Symbol: BIVM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204251.2 | NP_989582.1 | 9 - 10 | 1461 | 66 | |
| XM_015274245.2 | XP_015129731.1 | 10 - 11 | 1499 | 66 | ||
| GenBank mRNA | AF411387.2 | - | 9 - 10 | 1427 | 66 | |
| AF411388.2 | - | 10 - 11 | 1499 | 66 | ||
| CF256612.1 | - | 4 - 5 | 330 | 66 | ||
More Information
Set Membership: |
|

BIVM
Genomic Map