Assay Details
| Gene Symbol: PSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204811.2 | NP_990142.1 | 1 - 2 | 57 | 59 | |
| XM_015288193.2 | XP_015143679.1 | 1 - 2 | 69 | 59 | ||
| XM_015288194.2 | XP_015143680.1 | 1 - 2 | 69 | 59 | ||
| XM_015288195.2 | XP_015143681.1 | 1 - 2 | 69 | 59 | ||
| GenBank mRNA | AJ446376.1 | - | 1 - 2 | 66 | 59 | |
| AJ719872.1 | - | 1 - 2 | 66 | 59 | ||
More Information
Set Membership: |
|

PSAP
Genomic Map