Assay Details
| Gene Symbol: RGS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204842.2 | NP_990173.1 | 3 - 4 | 300 | 68 | |
| XM_015282661.1 | XP_015138147.1 | 3 - 4 | 361 | 68 | ||
| XM_025147032.1 | XP_025002800.1 | 3 - 4 | 1111 | 68 | ||
| GenBank mRNA | AF151967.2 | - | 3 - 4 | 300 | 68 | |
| MG576297.1 | - | 1 - 2 | 70 | 68 | ||
More Information
Set Membership: |
|

RGS20
Genomic Map