Assay Details
| Gene Symbol: FABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205308.2 | NP_990639.1 | 1 - 2 | 81 | 57 | |
| GenBank mRNA | X65459.1 | - | 81 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FABP7| Gene Symbol: FABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205308.2 | NP_990639.1 | 1 - 2 | 81 | 57 | |
| GenBank mRNA | X65459.1 | - | 81 | 57 | ||
Set Membership: |
|