Assay Details
| Gene Symbol: MDK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113289.1 | NP_001106760.1 | 1 - 2 | 78 | 55 | |
| XM_015287039.2 | XP_015142525.1 | 2 - 3 | 169 | 55 | ||
| XM_015287040.2 | XP_015142526.1 | 2 - 3 | 212 | 55 | ||
| XM_015287041.2 | XP_015142527.1 | 2 - 3 | 236 | 55 | ||
| GenBank mRNA | AF419846.1 | - | 2 - 3 | 187 | 55 | |
| M61754.1 | - | 1 - 2 | 105 | 55 | ||
More Information
Set Membership: |
|

MDK
Genomic Map