Assay Details
| Gene Symbol: PNPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113291.1 | NP_001106762.1 | 7 - 8 | 1110 | 64 | |
| XM_015286619.2 | XP_015142105.2 | 7 - 8 | 906 | 64 | ||
| GenBank mRNA | EU240627.2 | - | 7 - 8 | 1110 | 64 | |
More Information
Set Membership: |
|

PNPLA2
Genomic Map