Assay Details
Gene Symbol: CASP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113293.1 | NP_001106764.1 | 1 - 2 | 202 | 72 | |
XM_015272947.2 | XP_015128433.1 | 1 - 2 | 771 | 72 | ||
XM_015272948.2 | XP_015128434.1 | 1 - 2 | 771 | 72 | ||
GenBank mRNA | EU165361.1 | - | 1 - 2 | 202 | 72 | |
EU165362.1 | - | 1 - 2 | 202 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
