Assay Details
| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114081.1 | NP_001107553.1 | 1 | 848 | 121 | |
| XM_015284122.2 | XP_015139608.1 | 4 | 1114 | 121 | ||
| XM_015284123.2 | XP_015139609.1 | 4 | 1215 | 121 | ||
| GenBank mRNA | AM933593.1 | - | 1 | 848 | 121 | |
More Information
Set Membership: |
|

CCR6
Genomic Map