Assay Details
| Gene Symbol: MRPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114629.1 | NP_001108101.1 | 2 - 3 | 249 | 63 | |
| GenBank mRNA | AJ721057.1 | - | 2 - 3 | 249 | 63 | |
More Information
Set Membership: |
|

MRPL38
Genomic Map