Assay Details
| Gene Symbol: JMJD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114659.1 | NP_001108131.1 | 1 - 2 | 80 | 68 | |
| XM_025150712.1 | XP_025006480.1 | 1 - 2 | 2166 | 68 | ||
| GenBank mRNA | BX935903.2 | - | 1 - 2 | 80 | 68 | |
More Information
Set Membership: |
|

JMJD7
Genomic Map