Assay Details
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204119.1 | NP_989450.1 | 4 - 5 | 922 | 70 | |
GenBank mRNA | AF040634.1 | - | 4 - 5 | 864 | 70 | |
AF472618.1 | - | 4 - 5 | 922 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
