Assay Details
| Gene Symbol: RPL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204140.1 | NP_989471.1 | 3 - 4 | 254 | 84 | |
| XM_015299662.2 | XP_015155148.1 | 3 - 4 | 317 | 84 | ||
| GenBank mRNA | AB046396.1 | - | 3 - 4 | 254 | 84 | |
| CR338894.1 | - | 280 | 84 | |||
More Information
Set Membership: |
|

RPL36
Genomic Map