Assay Details
| Gene Symbol: CKMT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204182.2 | NP_989513.2 | 5 - 6 | 808 | 80 | |
| GenBank mRNA | BU118323.1 | - | 1 - 2 | 54 | 80 | |
| BU295295.1 | - | 5 - 6 | 663 | 80 | ||
More Information
Set Membership: |
|

CKMT1B
Genomic Map