Assay Details
| Gene Symbol: UNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204189.1 | NP_989520.1 | 1 - 2 | 96 | 66 | |
| GenBank mRNA | AF525946.1 | - | 1 - 2 | 96 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNG| Gene Symbol: UNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204189.1 | NP_989520.1 | 1 - 2 | 96 | 66 | |
| GenBank mRNA | AF525946.1 | - | 1 - 2 | 96 | 66 | |
Set Membership: |
|