Assay Details
| Gene Symbol: MYOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204214.2 | NP_989545.2 | 2 - 3 | 799 | 66 | |
| GenBank mRNA | FJ977569.1 | - | 2 - 3 | 777 | 66 | |
| X16189.1 | - | 796 | 66 | |||
More Information
Set Membership: |
|

MYOD1
Genomic Map