Assay Details
| Gene Symbol: CDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204227.1 | NP_989558.1 | 8 - 9 | 1470 | 56 | |
| XM_015292499.2 | XP_015147985.1 | 8 - 9 | 1528 | 56 | ||
| GenBank mRNA | AF522067.1 | - | 8 - 9 | 1470 | 56 | |
More Information
Set Membership: |
|

CDH5
Genomic Map