Assay Details
Gene Symbol: ERAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204239.2 | NP_989570.2 | 5 - 6 | 487 | 60 | |
GenBank mRNA | AB089163.1 | - | 5 - 6 | 484 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
