Assay Details
| Gene Symbol: RPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204273.1 | NP_989604.1 | 2 - 3 | 162 | 54 | |
| GenBank mRNA | AB046395.1 | - | 2 - 3 | 162 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL35| Gene Symbol: RPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204273.1 | NP_989604.1 | 2 - 3 | 162 | 54 | |
| GenBank mRNA | AB046395.1 | - | 2 - 3 | 162 | 54 | |
Set Membership: |
|