Assay Details
| Gene Symbol: ACTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204307.1 | NP_989638.1 | 6 - 7 | 621 | 85 | |
| XM_025152074.1 | XP_025007842.1 | 6 - 7 | 571 | 85 | ||
| GenBank mRNA | AF288600.1 | - | 6 - 7 | 621 | 85 | |
More Information
Set Membership: |
|

ACTR3
Genomic Map