Assay Details
Gene Symbol: POU1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204319.1 | NP_989650.1 | 5 - 6 | 853 | 67 | |
XM_015295651.2 | XP_015151137.1 | 6 - 7 | 2994 | 67 | ||
XM_015295656.2 | XP_015151142.1 | 6 - 7 | 2910 | 67 | ||
GenBank mRNA | AF029892.1 | - | 5 - 6 | 853 | 67 | |
AJ222692.2 | - | 5 - 6 | 769 | 67 | ||
AJ236855.1 | - | 6 - 7 | 793 | 67 | ||
AJ236856.1 | - | 6 - 7 | 877 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
