Assay Details
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305674.2 | NP_001292603.1 | 4 - 5 | 1006 | 59 | |
NM_204335.3 | NP_989666.1 | 4 - 5 | 1006 | 59 | ||
XM_015278173.2 | XP_015133659.1 | 7 - 8 | 3488 | 59 | ||
XM_015278174.2 | XP_015133660.1 | 6 - 7 | 1179 | 59 | ||
XM_015278175.2 | XP_015133661.1 | 5 - 6 | 1171 | 59 | ||
XM_015278176.2 | XP_015133662.1 | 5 - 6 | 1084 | 59 | ||
XM_015278177.2 | XP_015133663.1 | 7 - 8 | 3487 | 59 | ||
XM_015278178.2 | XP_015133664.1 | 7 - 8 | 3487 | 59 | ||
XM_015278179.2 | XP_015133665.1 | 7 - 8 | 3486 | 59 | ||
GenBank mRNA | AF330009.1 | - | 4 - 5 | 892 | 59 | |
CR406311.1 | - | 1 - 2 | 66 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
