Assay Details
| Gene Symbol: WNT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204336.2 | NP_989667.2 | 201 | 103 | ||
| GenBank mRNA | AF346628.1 | - | 161 | 103 | ||
| BX935899.1 | - | 202 | 103 | |||
More Information
Set Membership: |
|

WNT2B
Genomic Map