Assay Details
| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204379.2 | NP_989710.2 | 2 - 3 | 325 | 72 | |
| XM_015291626.2 | XP_015147112.1 | 3 - 4 | 696 | 72 | ||
| XM_015291627.2 | XP_015147113.1 | 3 - 4 | 692 | 72 | ||
| XM_015291628.2 | XP_015147114.1 | 2 - 3 | 298 | 72 | ||
| XM_015291629.2 | XP_015147115.1 | 3 - 4 | 620 | 72 | ||
| XM_015291630.2 | XP_015147116.1 | 3 - 4 | 620 | 72 | ||
| GenBank mRNA | AB114678.1 | - | 2 - 3 | 304 | 72 | |
| AJ720191.1 | - | 2 - 3 | 326 | 72 | ||
| AM066390.1 | - | 1 - 2 | 251 | 72 | ||
More Information
Set Membership: |
|

TNFSF10
Genomic Map