Assay Details
| Gene Symbol: PDE6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204408.1 | NP_989739.1 | 2 - 3 | 243 | 68 | |
| XM_015284399.2 | XP_015139885.1 | 2 - 3 | 585 | 68 | ||
| GenBank mRNA | AY226462.1 | - | 2 - 3 | 243 | 68 | |
More Information
Set Membership: |
|

PDE6H
Genomic Map