Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204430.2 | NP_989761.2 | 2 - 3 | 283 | 84 | |
| XM_015280410.2 | XP_015135896.1 | 2 - 3 | 370 | 84 | ||
| GenBank mRNA | AJ535692.1 | - | 2 - 3 | 285 | 84 | |
| BX929517.2 | - | 1 - 2 | 107 | 84 | ||
More Information
Set Membership: |
|

FUT10
Genomic Map