Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204467.1 | NP_989798.1 | 8 - 9 | 1109 | 76 | |
| GenBank mRNA | AY129071.1 | - | 8 - 9 | 1109 | 76 | |
| BU105681.1 | - | 3 - 4 | 327 | 76 | ||
More Information
Set Membership: |
|

RHCE
Genomic Map