Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204467.1 | NP_989798.1 | 9 - 10 | 1191 | 81 | |
| GenBank mRNA | AY129071.1 | - | 9 - 10 | 1191 | 81 | |
More Information
Set Membership: |
|

RHCE
Genomic Map