Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204507.1 | NP_989838.1 | 2 - 3 | 360 | 55 | |
| XM_015296153.2 | XP_015151639.1 | 2 - 3 | 376 | 55 | ||
| GenBank mRNA | AF373778.1 | - | 2 - 3 | 360 | 55 | |
| BX933105.1 | - | 2 - 3 | 239 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015296153.2 transcripts.
Set Membership: |
|

DHRS11
Genomic Map